Anti-NECTIN2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80327-RP01-400 80327-RP01-200 80327-RP01-100
103661-536EA 351.49 USD
103661-536 103661-538 103661-534
Anti-NECTIN2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat Nectin-2/PVRL2 (rR Nectin-2/PVRL2; Catalog#80327-R08H; Q9ESS6; Met1-Gly351). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: Nectin-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR