Anti-SEMA4D Rabbit Polyclonal Antibody

Supplier: Sino Biological

80320-T16-200 80320-T16-50 80320-T16-100
103597-214EA 214.31 USD
103597-214 103637-668 103597-212
Anti-SEMA4D Rabbit Polyclonal Antibody
Antibodies
Anti-Semaphorin 4D Antibody

Type: Primary
Antigen: SEMA4D
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR