Anti-SEMA4D Rabbit Polyclonal Antibody

Supplier: Sino Biological
80320-RP01-100 80320-RP01-200 80320-RP01-400
103661-528EA 351.49 USD
103661-528 103662-266 103662-268
Anti-SEMA4D Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat Semaphorin 4D/SEMA4D/CD100 (Catalog#80320-R08H; NP_001164034.1; Met1-Met712). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: SEMA4D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR