Anti-Tspan-30 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80304-T16-100 80304-T16-200 80304-T16-50
103597-176EA 160.46 USD
103597-176 103597-178 103637-870
Anti-Tspan-30 Rabbit Polyclonal Antibody
Antibodies
Anti-CD63 Antibody

Type: Primary
Antigen: Tspan-30
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR