Anti-CD59 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80299-T16-200 80299-T16-100 80299-T16-50
103637-658EA 142.96 USD
103637-658 103597-166 103597-164
Anti-CD59 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CD59 (Catalog#80299-R08H; P27274; Met1-Asn100). CD59 specific IgG was purified by Rat CD59 affinity chromatography.

Type: Primary
Antigen: CD59
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR