Anti-CD59 Rabbit Monoclonal Antibody [clone: 014]

Supplier: Sino Biological

80299-R014-50 80299-R014-100
103597-162EA 128.15 USD
103597-162 103661-484
Anti-CD59 Rabbit Monoclonal Antibody [clone: 014]
Antibodies
Anti-CD59 Antibody

Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: Clone: 014
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR