Anti-CD59 Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 001]

Supplier: Sino Biological

80299-R001-P-25 80299-R001-P-100
103630-246EA 32.31 USD
103630-246 103632-434
Anti-CD59 Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 001]
Antibodies
Anti-CD59 Antibody (PE)

Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: Clone: 001
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR