Anti-CD59 Rabbit Monoclonal Antibody [clone: 001]

Supplier: Sino Biological
80299-R001-100 80299-R001-50
103661-478EA 271.4 USD
103661-478 103597-156
Anti-CD59 Rabbit Monoclonal Antibody [clone: 001]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD59 (rR CD59; Catalog#80299-R08H; P27274; Met1-Asn100).

Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: 001
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR