Anti-CD48 Rabbit Monoclonal Antibody [clone: 001]

Supplier: Sino Biological
80286-R001-100 80286-R001-50
103661-468EA 271.4 USD
103661-468 103597-148
Anti-CD48 Rabbit Monoclonal Antibody [clone: 001]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD48/SLAMF2 (Catalog#80286-R08H; P10252; Met1-Arg216).

Type: Primary
Antigen: Anti-CD48 Antibody
Clonality: Monoclonal
Clone: 001
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR