Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDH17 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80283-T16-200 80283-T16-100 80283-T16-50
103597-146EA 214.31 USD InStock
103597-146 103597-144 103637-652
Anti-CDH17 Rabbit Polyclonal Antibody
Antibodies
Anti-CDH17 Antibody

Type: Primary
Antigen: CDH17
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty