Anti-CDH8 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80278-T24-100 80278-T24-50 80278-T24-200
103597-140EA 270.87 USD
103597-140 103637-650 103597-142
Anti-CDH8 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat Cadherin-8 / CDH8 (rR Cadherin-8 / CDH8; Catalog#80278-RCCH; NP_445845.2; Met1-Met621). Cadherin-8 / CDH8 specific IgG was purified by Rat Cadherin-8 / CDH8 affinity chromatography.

Type: Primary
Antigen: CDH8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR