Anti-CD144 Mouse Monoclonal Antibody [clone: 05]

Supplier: Sino Biological

80276-MM05-100 80276-MM05-200 80276-MM05-50
103661-454EA 160.46 USD
103661-454 103661-456 103597-128
Anti-CD144 Mouse Monoclonal Antibody [clone: 05]
Antibodies
Anti-VE-Cadherin Antibody

Type: Primary
Antigen: CD144
Clonality: Monoclonal
Clone: Clone: 05
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR