Anti-CDH1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80274-T50-100 80274-T50-50 80274-T50-200
103637-644EA 142.96 USD
103637-644 103597-126 103597-124
Anti-CDH1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat E-Cadherin / CDH1 / E-cad / CD324 (rR E-Cadherin / CDH1 / E-cad / CD324; Catalog#80274-R08H; Q9R0T4; Met1-Ala713). E-Cadherin / CDH1 / E-cad / CD324 specific IgG was purified by Rat E-Cadherin / CDH1 / E-cad / CD324 affinity chromatography.

Type: Primary
Antigen: CDH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR