Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DSC2 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80266-T24-50 80266-T24-200 80266-T24-100
103637-638EA 106.62 USD InStock
103637-638 103597-110 103597-108
Anti-DSC2 Rabbit Polyclonal Antibody
Antibodies
Anti-Desmocollin 2 Antibody

Type: Primary
Antigen: DSC2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty