Anti-DSC2 Rabbit Monoclonal Antibody [clone: 023]

Supplier: Sino Biological

80266-R023-50 80266-R023-100
103597-106EA 128.15 USD
103597-106 103661-422
Anti-DSC2 Rabbit Monoclonal Antibody [clone: 023]
Antibodies
Anti-Desmocollin 2 Antibody

Type: Primary
Antigen: DSC2
Clonality: Monoclonal
Clone: Clone: 023
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR