Anti-CD36 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80263-T48-100 80263-T48-50 80263-T48-200
103597-094EA 160.46 USD
103597-094 103638-050 103597-096
Anti-CD36 Rabbit Polyclonal Antibody
Antibodies
Anti-CD36 Antibody

Type: Primary
Antigen: CD36
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR