Anti-CD36 Rabbit Monoclonal Antibody [clone: 002]

Supplier: Sino Biological
80263-R002-100 80263-R002-50
103661-408EA 271.4 USD
103661-408 103597-092
Anti-CD36 Rabbit Monoclonal Antibody [clone: 002]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD36 / SCARB3 (rR CD36 / SCARB3; Catalog#80263-R08H; Q66HI4; Gly30-Lys439).

Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: 002
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR