Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CLEC5A Rabbit Polyclonal Antibody

Supplier: Sino Biological

80259-T16-100 80259-T16-200 80259-T16-50
103597-088EA 160.46 USD InStock
103597-088 103597-090 103638-048
Anti-CLEC5A Rabbit Polyclonal Antibody
Antibodies
Anti-CLEC5A Antibody

Type: Primary
Antigen: CLEC5A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty