Anti-CLEC5A Rabbit Polyclonal Antibody

Supplier: Sino Biological
80259-RP01-200 80259-RP01-100 80259-RP01-400
103661-404EA 351.49 USD
103661-404 103661-406 103661-402
Anti-CLEC5A Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CLEC5A / MDL1 / MDL-1 (Catalog#80259-R07H; F7GAV1; Val30-Lys190). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CLEC5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR