Anti-REG3A Rabbit Polyclonal Antibody

Supplier: Sino Biological
80258-T24-100 80258-T24-50 80258-T24-200
103597-084EA 213.93 USD
103597-084 103597-082 103638-046
Anti-REG3A Rabbit Polyclonal Antibody
Antibodies
Anti-Reg3A Antibody

Type: Primary
Antigen: REG3A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR