Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-REG3A Mouse Monoclonal Antibody [clone: 02]

Supplier: Sino Biological

80258-MM02-100 80258-MM02-200 80258-MM02-50
103662-260EA 160.46 USD InStock
103662-260 103662-262 103597-080
Anti-REG3A Mouse Monoclonal Antibody [clone: 02]
Antibodies
Anti-Reg3A Antibody

Type: Primary
Antigen: REG3A
Clonality: Monoclonal
Clone: Clone: 02
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty