Anti-Galectin 1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80248-RP01-400 80248-RP01-200 80248-RP01-100
103662-258EA 214.31 USD
103662-258 103662-256 103662-254
Anti-Galectin 1 Rabbit Polyclonal Antibody
Antibodies
Anti-Galectin-1 Antibody

Type: Primary
Antigen: Galectin 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR