Anti-LGALS1 Mouse Monoclonal Antibody [clone: 06]

Supplier: Sino Biological
80248-MM06-100 80248-MM06-200 80248-MM06-50
103662-250EA 352.03 USD
103662-250 103662-248 103597-068
Anti-LGALS1 Mouse Monoclonal Antibody [clone: 06]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat Galectin-1 / LGALS1 (rR Galectin-1 / LGALS1; P11762; Ala2-Glu135). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: Galectin 1
Clonality: Monoclonal
Clone: 06
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR