Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PVRL1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80244-T16-200 80244-T16-50 80244-T16-100
103597-066EA 214.31 USD InStock
103597-066 103637-632 103597-064
Anti-PVRL1 Rabbit Polyclonal Antibody
Antibodies
Anti-PVRL1 Antibody

Type: Primary
Antigen: PVRL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty