Anti-IL3 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80235-RP02-100 80235-RP02-200 80235-RP02-50
103661-394EA 160.46 USD
103661-394 103661-396 103597-054
Anti-IL3 Rabbit Polyclonal Antibody
Antibodies
Anti-IL3 Antibody

Type: Primary
Antigen: IL3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR