Anti-IL3 Rabbit Monoclonal Antibody [clone: 003]

Supplier: Sino Biological

80235-R003-50 80235-R003-100
103597-052EA 128.15 USD
103597-052 103662-234
Anti-IL3 Rabbit Monoclonal Antibody [clone: 003]
Antibodies
Anti-IL3 Antibody

Type: Primary
Antigen: IL3
Clonality: Monoclonal
Clone: Clone: 003
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR