Anti-IL3 Mouse Monoclonal Antibody [clone: 03]

Supplier: Sino Biological
80235-MM03-50 80235-MM03-100 80235-MM03-200
103661-392EA 352.03 USD
103661-392 103597-050 103661-390
Anti-IL3 Mouse Monoclonal Antibody [clone: 03]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat IL3 / IL-3 / Interleukin-3 (rR IL3 / IL-3 / Interleukin-3; Catalog#80235-R08B; NP_113701.1; Met1-Cys169). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IL3
Clonality: Monoclonal
Clone: 03
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR