Anti-CD38 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80229-T26-50 80229-T26-100 80229-T26-200
103637-624EA 142.96 USD
103637-624 103597-044 103597-046
Anti-CD38 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CD38/ADPRC1 (rR CD38/ADPRC1; Catalog#80229-R08H; Q64244; Trp45-Val303). CD38/ADPRC1 specific IgG was purified by Rat CD38/ADPRC1 affinity chromatography.

Type: Primary
Antigen: CD38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR