Anti-KIRREL3 Mouse Monoclonal Antibody [clone: 04]

Supplier: Sino Biological
80225-MM04-100 80225-MM04-200 80225-MM04-50
103661-370EA 270.87 USD
103661-370 103661-372 103597-038
Anti-KIRREL3 Mouse Monoclonal Antibody [clone: 04]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat KIRREL3/NEPH2 (Catalog#80225-R08H; Q09GS6; Met1-Ala523). The IgG fraction of the cell culture supernatant was purified by Protein A affinity.

Type: Primary
Antigen: KIRREL3
Clonality: Monoclonal
Clone: 04
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR