Anti-CEACAM1 Mouse Monoclonal Antibody [clone: 03]

Supplier: Sino Biological
80224-MM03-100 80224-MM03-200 80224-MM03-50
103597-030EA 142.96 USD
103597-030 103661-356 103661-358
Anti-CEACAM1 Mouse Monoclonal Antibody [clone: 03]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat CEACAM‑1/CD66a (rR CEACAM‑1/CD66a; Catalog#80224-R08H; Met1-Ser422). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CEACAM1
Clonality: Monoclonal
Clone: 03
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR