Anti-BCAM Rabbit Polyclonal Antibody

Supplier: Sino Biological
80222-RP02-200 80222-RP02-50 80222-RP02-100
103662-624EA 352.03 USD
103662-624 103597-028 103662-232
Anti-BCAM Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat BCAM (rR BCAM; Catalog#80222-R08H; Q9ESS6; Met1-Ala543). BCAM specific IgG was purified by Rat BCAM affinity chromatography.

Type: Primary
Antigen: BCAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR