Anti-BCAM Rabbit Polyclonal Antibody

Supplier: Sino Biological

80222-RP01-200 80222-RP01-400 80222-RP01-100
103662-228EA 160.46 USD
103662-228 103662-230 103662-226
Anti-BCAM Rabbit Polyclonal Antibody
Antibodies
Anti-BCAM Antibody

Type: Primary
Antigen: BCAM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR