Anti-BCAM Rabbit Monoclonal Antibody [clone: 005]

Supplier: Sino Biological
80222-R005-50 80222-R005-100
103597-024EA 150.18 USD
103597-024 103662-620
Anti-BCAM Rabbit Monoclonal Antibody [clone: 005]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat BCAM / CD239 (rR BCAM / CD239; Catalog#80222-R08H; Q9ESS6; Met1-Ala543).

Type: Primary
Antigen: BCAM
Clonality: Monoclonal
Clone: 005
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR