Anti-CD93 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80207-T24-200 80207-T24-100 80207-T24-50
103597-018EA 214.31 USD
103597-018 103597-016 103637-618
Anti-CD93 Rabbit Polyclonal Antibody
Antibodies
Anti-CD93 Antibody

Type: Primary
Antigen: CD93
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR