Anti-CD93 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80207-RP01-200 80207-RP01-400 80207-RP01-100
103661-340EA 351.49 USD
103661-340 103661-342 103661-344
Anti-CD93 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CD93 / C1qR (Catalog#80207-R08H; Q9ET61; Met1-Lys571). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CD93
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR