Anti-CD93 Rabbit Monoclonal Antibody [clone: 138] FITC (Fluorescein Isothiocyanate)

Supplier: Sino Biological
80207-R138-F-25 80207-R138-F-100
103630-232EA 90.9 USD
103630-232 103632-412
Anti-CD93 Rabbit Monoclonal Antibody [clone: 138] FITC (Fluorescein Isothiocyanate)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD93/C1qR (rR CD93/C1qR; Catalog#80207-R08H; Q9ET61; Met1-Lys571) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: CD93
Clonality: Monoclonal
Clone: Clone: 138
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR