Anti-CD93 Rabbit Monoclonal Antibody [clone: 138]

Supplier: Sino Biological
80207-R138-50 80207-R138-100
103597-006EA 150.18 USD
103597-006 103661-334
Anti-CD93 Rabbit Monoclonal Antibody [clone: 138]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD93/C1qR (rR CD93/C1qR; Catalog#80207-R08H; Q9ET61; Met1-Lys571).

Type: Primary
Antigen: CD93
Clonality: Monoclonal
Clone: 138
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR