Anti-IFNA5 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80205-T60-200 80205-T60-50 80205-T60-100
103637-612EA 142.96 USD
103637-612 103597-004 103597-002
Anti-IFNA5 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat IFNA5 / IFNaG (Catalog#80205-R08H; XP_001076062.1; Met1-Glu189). IFNA5 / IFNaG specific IgG was purified by Rat IFNA5 / IFNaG affinity chromatography.

Type: Primary
Antigen: IFNA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR