Anti-IFNA5 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80205-RP01-200 80205-RP01-100 80205-RP01-400
103661-328EA 160.46 USD
103661-328 103661-330 103661-332
Anti-IFNA5 Rabbit Polyclonal Antibody
Antibodies
Anti-interferon-alpha Antibody

Type: Primary
Antigen: IFNA5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR