Anti-IL13Ra2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80204-RP01-400 80204-RP01-100 80204-RP01-200
103661-316EA 270.2 USD
103661-316 103661-318 103661-320
Anti-IL13Ra2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat IL13RA2/IL13R (rR IL13RA2/IL13R; Catalog#80204-R08H; Q8VHK6; Met1-Lys336). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IL13RA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR