Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL13RA2 Rabbit Monoclonal Antibody [clone: 007]

Supplier: Sino Biological

80204-R007-50 80204-R007-100
103596-996EA 128.15 USD InStock
103596-996 103662-616
Anti-IL13RA2 Rabbit Monoclonal Antibody [clone: 007]
Antibodies
Anti-IL13RA2 Antibody

Type: Primary
Antigen: IL13RA2
Clonality: Monoclonal
Clone: Clone: 007
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty