Anti-IL4R Rabbit Polyclonal Antibody

Supplier: Sino Biological
80198-T16-50 80198-T16-100 80198-T16-200
103637-608EA 142.96 USD
103637-608 103596-986 103596-988
Anti-IL4R Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat IL-4R / CD124 (rR IL-4R / CD124; Catalog#80198-R08H; Q63257-1; Met1-Arg232). IL-4R / CD124 specific IgG was purified by Rat IL-4R / CD124 affinity chromatography.

Type: Primary
Antigen: IL4R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR