Anti-IL4R Rabbit Polyclonal Antibody

Supplier: Sino Biological
80198-RP01-400 80198-RP01-100 80198-RP01-200
103661-308EA 351.49 USD
103661-308 103661-312 103661-310
Anti-IL4R Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat IL-4R / CD124 (rR IL-4R / CD124; Catalog#80198-R08H; Q63257-1; Met1-Arg232). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IL4R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR