Anti-IL2RG Rabbit Polyclonal Antibody

Supplier: Sino Biological
80197-T16-50 80197-T16-200 80197-T16-100
103637-606EA 142.96 USD
103637-606 103596-982 103596-980
Anti-IL2RG Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CD132/IL2RG (Catalog#80197-R08H; Q68FU6; Met1-Ala262). CD132/IL2RG specific IgG was purified by Rat CD132/IL2RG affinity chromatography.

Type: Primary
Antigen: CD132
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR