Anti-IL1R2 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80196-RP01-100 80196-RP01-200 80196-RP01-400
103661-304EA 106.62 USD
103661-304 103661-302 103662-218
Anti-IL1R2 Rabbit Polyclonal Antibody
Antibodies
Anti-IL1R2 Antibody

Type: Primary
Antigen: IL1R2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR