Anti-IL1R2 Rabbit Monoclonal Antibody [clone: 161]

Supplier: Sino Biological
80196-R161-100 80196-R161-1 80196-R161-50
103637-604EA 1500.59 USD
103637-604 103596-978 103661-300
Anti-IL1R2 Rabbit Monoclonal Antibody [clone: 161]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL-1R2/CD121b (rR IL-1R2/CD121b; Catalog#80196-R08H; P43303; Met1-Glu355).

Type: Primary
Antigen: IL1R2
Clonality: Monoclonal
Clone: 161
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR