Anti-IL17RA Rabbit Polyclonal Antibody

Supplier: Sino Biological
80190-T16-200 80190-T16-50 80190-T16-100
103596-972EA 352.03 USD
103596-972 103637-600 103596-970
Anti-IL17RA Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat IL-17R / IL-17RA (CD217) (Catalog#80190-R08H; NP_001101353.2; Met1-Pro320). IL-17R / IL-17RA (CD217) specific IgG was purified by Rat IL-17R / IL-17RA (CD217) affinity chromatography.

Type: Primary
Antigen: IL17RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR