Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL17RA Mouse Monoclonal Antibody [clone: 05]

Supplier: Sino Biological

80190-MM05-100 80190-MM05-50 80190-MM05-200
103662-602EA 160.46 USD InStock
103662-602 103596-968 103662-604
Anti-IL17RA Mouse Monoclonal Antibody [clone: 05]
Antibodies
Anti-IL17RA Antibody

Type: Primary
Antigen: IL17RA
Clonality: Monoclonal
Clone: Clone: 05
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty