Anti-IL20RB Rabbit Polyclonal Antibody

Supplier: Sino Biological

80187-RP01-200 80187-RP01-400 80187-RP01-100
103662-598EA 160.46 USD
103662-598 103662-600 103662-596
Anti-IL20RB Rabbit Polyclonal Antibody
Antibodies
Anti-IL20RB Antibody

Type: Primary
Antigen: IL20RB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR