Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL20RB Rabbit Monoclonal Antibody [clone: 022]

Supplier: Sino Biological

80187-R022-100 80187-R022-50
103662-594EA 214.31 USD InStock
103662-594 103596-958
Anti-IL20RB Rabbit Monoclonal Antibody [clone: 022]
Antibodies
Anti-IL20RB Antibody

Type: Primary
Antigen: IL20RB
Clonality: Monoclonal
Clone: Clone: 022
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty